Benzyl-4-chlorophenyl ketone ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A15959.06 A15959.14 A15959.22
A15959.06EA 45.1 EUR
A15959.06 A15959.14 A15959.22
Benzyl-4-chlorophenyl ketone ≥98%
Benzyl-4-chlorophenyl ketone

Purity: 98% CAS No.: 1889-71-0 Molecular Formula: C14H11ClO Formula Weight: 230.69 Melting Point: 103-107° Beilstein Reference: 976443

Formule: C₁₄H₁₁ClO
Smeltpunt: 103…107 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016342
CAS nummer: 1889-71-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR