4-Amino-N,N-dimethylaniline 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A15962.0B A15962.18 A15962.30
A15962.0BEA 572 EUR
A15962.0B A15962.18 A15962.30
4-Amino-N,N-dimethylaniline 96%
4-Amino-N,N-dimethylaniline

Purity: 96% CAS No.: 99-98-9 Molecular Formula: C8H12N2 Formula Weight: 136.20 Flash Point: 130°(266°F) Melting Point: 34-38° Boiling Point: 262° Beilstein Reference: 508105 Merck Reference: 14,3254 UN No.: 2811 MDL No.: MFCD00007860

Formule: C₈H₁₂N₂
Kookpunt: 262 °C
Smeltpunt: 34…38 °C
Dichtheid: 1,09
Vlampunt: 130 °C (266 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007860
CAS nummer: 99-98-9
EINECS: 202-807-5
UN: 2811
ADR: 6.1,II
Merck Index: 14,3254

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR