1,8-Naftyleendiamine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A15966.18 A15966.0B A15966.30
A15966.18EA 49.2 EUR
A15966.18 A15966.0B A15966.30
1,8-Naftyleendiamine 97%
1,8-Naftyleendiamine

Purity: 97% CAS No.: 479-27-6 Molecular Formula: C10H10N2 Formula Weight: 158.20 Flash Point: 195°(383°F) Melting Point: 61-65° Boiling Point: 205°/12mm Beilstein Reference: 742841 Merck Reference: 14,6372 MDL No.: MFCD00004033

Formule: C₁₀H₁₀N₂
Kookpunt: 205 °C (12 mmHg)
Smeltpunt: 61…65 °C
Dichtheid: 1,18 g/cm³ (20 °C)
Vlampunt: 195 °C (383 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004033
CAS nummer: 479-27-6
EINECS: 207-529-8
UN: 3259
ADR: 8,III
Merck Index: 14,6372

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR