1,3-Dimethyl-2-imidazolidinone ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1,3-Dimethylimidazolidin-2-one

A16001.22 A16001.14 A16001.36
A16001.22EA 55.2 EUR
A16001.22 A16001.14 A16001.36
1,3-Dimethyl-2-imidazolidinone ≥98%
1,3-Dimethyl-2-imidazolidinone

Purity: 98% CAS No.: 80-73-9 Molecular Formula: C5H10N2O Formula Weight: 114.15 Flash Point: 93°(199°F) Refractive Index: 1.4720 Melting Point: 8° Boiling Point: 224-226° Beilstein Reference: 108808 Merck Reference: 14,3249 UN No.: 2810 MDL No.: MFCD00003188

Formule: C₅H₁₀N₂O
Kookpunt: 224…226 °C
Smeltpunt: 8 °C
Dichtheid: 1,058
Vlampunt: 93 °C (199 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003188
CAS nummer: 80-73-9
EINECS: 201-304-8
UN: 2810
ADR: 6.1,III
Merck Index: 14,3249

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR