PIPES ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A16090.14 A16090.22 A16090.36
A16090.14EA 34.8 EUR
A16090.14 A16090.22 A16090.36
PIPES ≥98%
PIPES

Purity: 98% CAS No.: 5625-37-6 Molecular Formula: C8H18N2O6S2 Formula Weight: 302.37 Melting Point: 300° Beilstein Reference: 817713 Merck Reference: 14,7479 MDL No.: MFCD00006159

Formule: C₈H₁₈N₂O₆S₂
Kookpunt: 499 °C (1013 hPa)
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006159
CAS nummer: 5625-37-6
EINECS: 227-057-6
Merck Index: 14,7479
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5192468b680c7bea85a31a131811618356a531b9&vid=ef0e3d488c68ea7f6f583fede6ebf7d2196392b7&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR