di-Sodium tetraborate decahydrate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A16176.0C A16176.0E A16176.36
A16176.0CEA 267 EUR
A16176.0C A16176.0E A16176.36
di-Sodium tetraborate decahydrate ≥99%
di-Sodium tetraborate decahydrate

Purity: 99+% CAS No.: 1303-96-4 Molecular Formula: B4Na2O7.10H2O Formula Weight: 381.37 (201.22anhy) Odour: Odourless pH: 9,24 Melting Point: 75° Merck Reference: 14,8590 MDL No.: MFCD00149193

Formule: Na₂B₄O₇·10H₂O
Kookpunt: 320 °C (1013 hPa)
Smeltpunt: 75 °C
Dichtheid: 1,73
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149193
CAS nummer: 1303-96-4
EINECS: 215-540-4
Bereik: 01-2119490790-32
Merck Index: 14,8590

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR