4-(Methylamino)phenol sulphate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A16236.0B A16236.18 A16236.30
A16236.0BEA 193 EUR
A16236.0B A16236.18 A16236.30
4-(Methylamino)phenol sulphate 99%
4-(Methylamino)phenol sulphate

Purity: 99% CAS No.: 55-55-0 Molecular Formula: C14H18N2O2.H2SO4 Formula Weight: 344.38 Melting Point: ca 260° dec. Beilstein Reference: 3919382 Merck Reference: 14,6017 UN No.: 2811 MDL No.: MFCD00013140

Formule: C₁₄H₂₀N₂O₆S
Kookpunt: 227 °C (1013 hPa)
Dichtheid: 1,434 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013140
CAS nummer: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,6017

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR