D-(+)-Maltose monohydrate 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A16266.22 A16266.36
A16266.22EA 42.1 EUR
A16266.22 A16266.36
D-(+)-Maltose monohydrate 95%
D-(+)-Maltose monohydrate

Purity: 95% CAS No.: 6363-53-7 Molecular Formula: C12H22O11.H2O Formula Weight: 360.32 (342.30anhy) Melting Point: ca 120° dec. Solubility: Soluble in water. Slightly soluble in alcohol. Insoluble in ether Beilstein Reference: 5784659 Merck Reference: 14,5714 Optical Rotation: +130° (c=2 in water, 24h) MDL No.: MFCD00149343

Formule: C₁₂H₂₂O₁₁·1H₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149343
CAS nummer: 6363-53-7
EINECS: 200-716-5
Merck Index: 14,5714
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR