Barium bromide dihydrate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A16328.22 A16328.36
A16328.22EA 29.7 EUR
A16328.22 A16328.36
Barium bromide dihydrate ≥98%
Barium bromide dihydrate
Formule: BaBr₂·2H₂O
Smeltpunt: 850 °C
Dichtheid: 3,58
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149153
CAS nummer: 7791-28-8
EINECS: 234-140-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,968

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR