Propagyl bromide ≥80% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Stabilisatie: Stabiised with magnesium oxide (MgO)

A16580.AD A16580.AK A16580.AU
A16580.ADEA 48.6 EUR
A16580.AD A16580.AK A16580.AU
Propagyl bromide ≥80% stabilised
Propagyl bromide

CAS No.: 106-96-7 Molecular Formula: HC≡CCH2Br Formula Weight: 118.97 Storage Temperature: 2°C to 8°C Flash Point: 3°(37°F) Refractive Index: 1.4940 Melting Point: -61° Boiling Point: 88-90° Beilstein Reference: 605309 UN No.: 2924 MDL No.: MFCD00000241

Formule: C₃H₃Br
Kookpunt: 88…90 °C
Smeltpunt: –61 °C
Dichtheid: 1,335
Vlampunt: 3 °C (37 °F)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000241
CAS nummer: 106-96-7
EINECS: 203-447-1
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR