Natriumchloride 0,9% in aqueous solution, sterile

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Saline, NaCl, Sea salt, Halite

A1671.0100 A1671.0250 A1671.0500 A1671.1000
A1671.0100EA 49.7 EUR
A1671.0100 A1671.0250 A1671.0500 A1671.1000
Natriumchloride 0,9% in aqueous solution, sterile
Natriumchloride
Formule: NaCl
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003477
CAS nummer: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

pH (20 °C) 5,0 - 7,5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR