(1-Bromoethyl)benzene ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A16839.14 A16839.22 A16839.36
A16839.14EA 53.2 EUR
A16839.14 A16839.22 A16839.36
(1-Bromoethyl)benzene ≥97%
(1-Bromoethyl)benzene

Purity: 97% CAS No.: 585-71-7 Molecular Formula: C8H9Br Formula Weight: 185.06 Flash Point: 81°(177°F) Refractive Index: 1.5600 Melting Point: -65° Boiling Point: 200° Beilstein Reference: 507210 MDL No.: MFCD00000139

Formule: C₆H₅CHBrCH₃
Kookpunt: 200 °C
Smeltpunt: –65 °C
Dichtheid: 1,356
Vlampunt: 81 °C (177 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000139
CAS nummer: 585-71-7
EINECS: 209-560-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f329f7855ae4dffcd097f49d6d504b1ec165e692&vid=6d612184698aeb1969948bdeb536d2ee39ec9551&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR