Benzyltrimethylammonium bromide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A16840.14 A16840.22
A16840.14EA 22.7 EUR
A16840.14 A16840.22
Benzyltrimethylammonium bromide ≥98%
Benzyltrimethylammonium bromide

Purity: 98+% CAS No.: 5350-41-4 Molecular Formula: C6H5CH2N(CH3)3Br Formula Weight: 230.15 Melting Point: 228-232° Beilstein Reference: 3917251

Formule: C₁₀H₁₆BrN
Smeltpunt: 228…232 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011780
CAS nummer: 5350-41-4
EINECS: 226-320-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR