Rose Bengal

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Sodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Food Red No. 105, CI 45440, Acid red 94, Aniline Rose, Rose Bengal sodium

A17053.14 A17053.22 A17053.36
A17053.14EA 41 EUR
A17053.14 A17053.22 A17053.36
Rose Bengal
Rose Bengal

CAS No.: 632-69-9 Molecular Formula: C20H2Cl4I4Na2O5 Formula Weight: 1017.65 Melting Point: 300° Beilstein Reference: 3645857 MDL No.: MFCD00151169

Formule: C₂₀H₂Cl₄I₄O₅Na₂
MW: 1017.63 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00151169
CAS nummer: 632-69-9
EINECS: 211-183-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR