3-Indoxyl sulphate potassium salt 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A17079.03 A17079.01 A17079.06
A17079.03EA 195 EUR
A17079.03 A17079.01 A17079.06
3-Indoxyl sulphate potassium salt 97%
3-Indoxyl sulphate potassium salt

Purity: 97% CAS No.: 2642-37-7 Molecular Formula: C8H6KNO4S Formula Weight: 251.31 Storage Temperature: -30°C to -10°C Melting Point: 165° dec. Beilstein Reference: 3921324 MDL No.: MFCD00037931

Formule: C₈H₆KNO₄S
MDL Nummer: MFCD00037931
CAS nummer: 2642-37-7
EINECS: 220-145-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR