Aluminiumoxid geactiveerd neutrale Brockmann Grade II

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A17141.0E A17141.36
A17141.0EEA 164 EUR
A17141.0E A17141.36
Aluminiumoxid geactiveerd neutrale Brockmann Grade II
Aluminium oxide activated neutral

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Flash Point: None Odour: Odourless MDL No.: MFCD00003424

Formule: Al₂O₃
Dichtheid: 3,97
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR