α,α'-Dibromo-o-xylene ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A17241.14 A17241.06 A17241.22
A17241.14EA 52 EUR
A17241.14 A17241.06 A17241.22
α,α'-Dibromo-o-xylene ≥97%
α,α'-Dibromo-o-xylene

Purity: 97% CAS No.: 91-13-4 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 92-96° Beilstein Reference: 637159 UN No.: 3261 MDL No.: MFCD00000175

Formule: C₈H₈Br₂
Kookpunt: 140 °C (20 mmHg)
Smeltpunt: 92…96 °C
Dichtheid: 1,960
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000175
CAS nummer: 91-13-4
EINECS: 202-042-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR