Methyl trifluoroacetate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Trifluoroacetic methyl ester

A17338.0B A17338.18 A17338.30
A17338.0BEA 636 EUR
A17338.0B A17338.18 A17338.30
Methyl trifluoroacetate ≥98%
Methyl trifluoroacetate

Purity: 98+% CAS No.: 431-47-0 Molecular Formula: CF3CO2CH3 Formula Weight: 128.05 Flash Point: -18°(-0.4°F) Refractive Index: 1.2900 Odour: Pungent Melting Point: -78° Boiling Point: 43-44° Beilstein Reference: 1756070 UN No.: 2924 MDL No.: MFCD00000417

Formule: C₃H₃F₃O₂
MW: 128.05 g/mol
Kookpunt: 43…44 °C
Smeltpunt: –78 °C
Dichtheid: 1.283
Vlampunt: –18 °C (–0.4 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000417
CAS nummer: 431-47-0
EINECS: 207-074-5
UN: 2924
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR