2-Bromethylamine hydrobromide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

A17625.18 A17625.0B A17625.30
A17625.18EA 26.5 EUR
A17625.18 A17625.0B A17625.30
2-Bromethylamine hydrobromide ≥98%
2-Bromethylamine hydrobromide

Purity: 98+% CAS No.: 2576-47-8 Molecular Formula: BrCH2CH2NH2.HBr Formula Weight: 204.90 Melting Point: 172-176° Beilstein Reference: 3607605

Formule: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Smeltpunt: 172…176 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012886
CAS nummer: 2576-47-8
EINECS: 219-924-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR