Menthone (mixture of isomers) ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A17665.0B A17665.18 A17665.30
A17665.0BEA 365 EUR
A17665.0B A17665.18 A17665.30
Menthone (mixture of isomers) ≥98%
Menthone (mixture of isomers)

Purity: 98% CAS No.: 10458-14-7 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 72°(162°F) Boiling Point: ca 207° MDL No.: MFCD00062998

Formule: C₁₀H₁₈O
Kookpunt: ∼207 °C
Dichtheid: 0,894
Vlampunt: 72 °C (162 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00062998
CAS nummer: 10458-14-7
EINECS: 233-944-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR