D-(+)-Tyrosine ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A17709.03 A17709.06 A17709.14
A17709.03EA 29.2 EUR
A17709.03 A17709.06 A17709.14
D-(+)-Tyrosine ≥99%
D-(+)-Tyrosine

Purity: 99% CAS No.: 556-02-5 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Melting Point: 310-314° Beilstein Reference: 2212157 Merck Reference: 14,9839 Optical Rotation: +11° (c=4 in 1N HCl) MDL No.: MFCD00063073

Formule: C₉H₁₁NO₃
Kookpunt: 351 °C (1013 hPa)
Smeltpunt: 310…314 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063073
CAS nummer: 556-02-5
EINECS: 209-112-6
Merck Index: 14,9839

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR