Gibberellic acid ≥90%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A17843.03 A17843.06 A17843.14
A17843.03EA 39.3 EUR
A17843.03 A17843.06 A17843.14
Gibberellic acid ≥90%
Gibberellic acid

Purity: 90+% CAS No.: 77-06-5 Molecular Formula: C19H22O6 Formula Weight: 346.37 Melting Point: 222° dec. Beilstein Reference: 54346 Merck Reference: 14,4419 Optical Rotation: +78° (c=2 in methanol) MDL No.: MFCD00079329

Formule: C₁₉H₂₂O₆
Dichtheid: 1,386 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00079329
CAS nummer: 77-06-5
EINECS: 201-001-0
Merck Index: 14,4419

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR