DL-Malic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A17874.0E A17874.30 A17874.36
A17874.0EEA 70.1 EUR
A17874.0E A17874.30 A17874.36
DL-Malic acid ≥98%
DL-Malic acid

Purity: 98% CAS No.: 6915-15-7 Molecular Formula: C4H6O5 Formula Weight: 134.09 Flash Point: 203°(397°F) Physical Form: Powder Density: 1.609 Appearance: White to pale cream Melting Point: 130-133° Solubility: Soluble in acetone, dioxane, water, methanol and ethanol. Insoluble in benzene. Beilstein Reference: 1723539 Merck Reference: 14,5707 MDL No.: MFCD00064212

Formule: HOOCCH(OH)CH₂COOH
Kookpunt: 306 °C (1013 hPa)
Smeltpunt: 130…133 °C
Dichtheid: 1,609
Vlampunt: 203 °C (397 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064212
CAS nummer: 617-48-1
EINECS: 230-022-8
Merck Index: 14,5707
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR