6-Methyl-2-thiouracil ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A17982.18 A17982.30 A17982.A1
A17982.18EA 20.4 EUR
A17982.18 A17982.30 A17982.A1
6-Methyl-2-thiouracil ≥98%
6-Methyl-2-thiouracil

Purity: 98% CAS No.: 56-04-2 Molecular Formula: C5H6N2OS Formula Weight: 142.18 Melting Point: ca 329° dec. Beilstein Reference: 115648 Merck Reference: 14,6128 MDL No.: MFCD00006040

Formule: C₅H₆N₂OS
Kookpunt: 351 °C (1013 hPa)
Dichtheid: 1,519 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006040
CAS nummer: 56-04-2
EINECS: 200-252-3
Merck Index: 14,6128

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR