Isonicotinic acid ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A18109.0E A18109.22 A18109.36
A18109.0EEA 460 EUR
A18109.0E A18109.22 A18109.36
Isonicotinic acid ≥99%
Isonicotinic acid

Purity: 99% CAS No.: 55-22-1 Molecular Formula: C6H5NO2 Formula Weight: 123.11 pH: 3.0-4.0 Melting Point: ca 310° subl. Boiling Point: 260°/15mm Beilstein Reference: 109599 Merck Reference: 14,5187 MDL No.: MFCD00006429

Formule: C₆H₅NO₂
Kookpunt: 260 °C (15 mmHg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006429
CAS nummer: 55-22-1
EINECS: 200-228-2
Merck Index: 14,5187

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR