Isopropyl myristate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A18111.AP A18111.AE A18111.0F
A18111.APEA 85.9 EUR
A18111.AP A18111.AE A18111.0F
Isopropyl myristate ≥98%
Isopropyl myristate

Purity: 98% CAS No.: 110-27-0 Molecular Formula: CH3(CH2)12CO2CH(CH3)2 Formula Weight: 270.46 Flash Point: 152°(306°F) Physical Form: Liquid Density: 0.85 Appearance: Colourless Boiling Point: 191-194°/20mm MDL No.: MFCD00008982

Formule: C₁₇H₃₄O₂
Kookpunt: 191…194 °C (20 mmHg)
Smeltpunt: 0…1 °C
Dichtheid: 0,85
Vlampunt: 152 °C (306 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008982
CAS nummer: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR