Phenyl propargyl ether ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A18272.03 A18272.06 A18272.14
A18272.03EA 37.8 EUR
A18272.03 A18272.06 A18272.14
Phenyl propargyl ether ≥98%
Phenyl propargyl ether

Purity: 98+% CAS No.: 13610-02-1 Molecular Formula: C9H8O Formula Weight: 132.16 Storage Temperature: 2°C to 8°C Flash Point: 76°(169°F) Refractive Index: 1.5330 Odour: Pungent Boiling Point: 89-90°/14mm MDL No.: MFCD00048107

Formule: C₉H₈O
Kookpunt: 89…90 °C (14 mmHg)
Dichtheid: 1,030
Vlampunt: 76 °C (169 °F)
MDL Nummer: MFCD00048107
CAS nummer: 13610-02-1
EINECS: 237-095-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR