1,4-Diaminobutane dihydrochloride 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A18312.22 A18312.14
A18312.22EA 191 EUR
A18312.22 A18312.14
1,4-Diaminobutane dihydrochloride 99%
1,4-Diaminobutane dihydrochloride

Purity: 99% CAS No.: 333-93-7 Molecular Formula: HCl.H2N(CH2)4NH2.HCl Formula Weight: 161.08 Melting Point: ca 280° dec. Beilstein Reference: 3906680 Merck Reference: 14,7947 MDL No.: MFCD00012526

Formule: C₄H₁₄Cl₂N₂
Dichtheid: 1,31 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012526
CAS nummer: 333-93-7
EINECS: 206-375-9
Merck Index: 14,7947

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR