2-Bromobenzamide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A18331.06 A18331.14
A18331.06EA 57 EUR
A18331.06 A18331.14
2-Bromobenzamide ≥98%
2-Bromobenzamide

Purity: 98% CAS No.: 4001-73-4 Molecular Formula: C7H6BrNO Formula Weight: 200.03 Melting Point: 160-162° MDL No.: MFCD00007969

Formule: C₇H₆BrNO
MW: 200.03 g/mol
Kookpunt: 198
Smeltpunt: 160…162 °C
Dichtheid: 1.609 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007969
CAS nummer: 4001-73-4
EINECS: 223-650-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR