O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A18368.01 A18368.03 A18368.06
A18368.01EA 62.2 EUR
A18368.01 A18368.03 A18368.06
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥99%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride

Purity: 99+% CAS No.: 57981-02-9 Molecular Formula: C7H4F5NO.HCl Formula Weight: 249.57 Melting Point: ca 215° subl. Beilstein Reference: 4031190 MDL No.: MFCD00012953

Formule: C₇H₄F₅NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012953
CAS nummer: 57981-02-9
EINECS: 261-057-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR