4,4'-(Hexafluoroisopropylidene)diphenol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A18370.06 A18370.14 A18370.22
A18370.06EA 32.5 EUR
A18370.06 A18370.14 A18370.22
4,4'-(Hexafluoroisopropylidene)diphenol ≥98%
4,4'-(Hexafluoroisopropylidene)diphenol

Purity: 98% CAS No.: 1478-61-1 Molecular Formula: C15H10F6O2 Formula Weight: 336.24 Melting Point: 158-162° Boiling Point: 400° MDL No.: MFCD00000439

Formule: C₁₅H₁₀F₆O₂
Kookpunt: 400 °C
Smeltpunt: 158…162 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000439
CAS nummer: 1478-61-1
EINECS: 216-036-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR