Streptomycin sulphate

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A1852.0025 A1852.1000
A1852.0025EA 37.4 EUR
A1852.0025 A1852.1000
Streptomycin sulphate
Streptomycin sulphate
Formule: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Smeltpunt: 250 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00037023
CAS nummer: 3810-74-0
EINECS: 223-286-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,08983

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR