β-Alanine benzyl ester p-toluenesulphonate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A18650.09 A18650.18
A18650.09EA 35 EUR
A18650.09 A18650.18
β-Alanine benzyl ester p-toluenesulphonate 98%
β-Alanine benzyl ester p-toluenesulphonate

Purity: 98% CAS No.: 27019-47-2 Molecular Formula: C6H5CH2O2CCH2CH2NH2.HO3SC6H4CH3 Formula Weight: 351.42 Storage Temperature: 2°C to 8°C Melting Point: 138-139° MDL No.: MFCD00039061

Formule: C₁₀H₁₃NO₂
Smeltpunt: 138…139 °C
MDL Nummer: MFCD00039061
CAS nummer: 27019-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR