3-Bromo-4-hydroxybenzonitrile ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A19400.06 A19400.14
A19400.06EA 46 EUR
A19400.06 A19400.14
3-Bromo-4-hydroxybenzonitrile ≥98%
3-Bromo-4-hydroxybenzonitrile

Purity: 98% CAS No.: 2315-86-8 Molecular Formula: C7H4BrNO Formula Weight: 198.02 Melting Point: 156-161° Beilstein Reference: 2207020 UN No.: 3439 MDL No.: MFCD00143096

Formule: C₇H₄BrNO
Kookpunt: 406 °C (1013 hPa)
Smeltpunt: 156…161 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00143096
CAS nummer: 2315-86-8
EINECS: 219-022-9
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR