Lead(II) bromide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Lead dibromide, Lead(II) bromide

A19406.22 A19406.36
A19406.22EA 30.2 EUR
A19406.22 A19406.36
Lead(II) bromide ≥98%
Lead(II) bromide

Purity: 98+% CAS No.: 10031-22-8 Molecular Formula: Br2Pb Formula Weight: 367.01 Odour: Odourless Melting Point: 373° Boiling Point: 916° Merck Reference: 14,5403 UN No.: 2291 MDL No.: MFCD00011156

Formule: Br₂Pb
MW: 367.01 g/mol
Kookpunt: 916 °C
Smeltpunt: 373 °C
Dichtheid: 6.66
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011156
CAS nummer: 10031-22-8
EINECS: 233-084-4
UN: 2291
ADR: 6.1 III
Merck Index: 12,05418

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR