(4-(Trifluoromethoxy)phenyl)acetic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A19512.03 A19512.06
A19512.03EA 55.5 EUR
A19512.03 A19512.06
(4-(Trifluoromethoxy)phenyl)acetic acid ≥98%
(4-(Trifluoromethoxy)phenyl)acetic acid

Purity: 98% CAS No.: 4315-07-5 Molecular Formula: C9H7F3O3 Formula Weight: 220.15 Melting Point: 86-88° Beilstein Reference: 2695336

Formule: (C₆H₄(OCF₃))CH₂COOH
Smeltpunt: 86…88 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00061239
CAS nummer: 4315-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR