1,3-Dimethyladamantane ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A19684.14 A19684.06
A19684.14EA 130 EUR
A19684.14 A19684.06
1,3-Dimethyladamantane ≥98%
1,3-Dimethyladamantane

Purity: 98% CAS No.: 702-79-4 Molecular Formula: C12H20 Formula Weight: 164.29 Flash Point: 52°(126°F) Refractive Index: 1.4780 Boiling Point: 201° UN No.: 3295 MDL No.: MFCD00074755

Formule: C₁₂H₂₀
Kookpunt: 201 °C
Smeltpunt: –30 °C
Dichtheid: 0,902
Vlampunt: 52 °C (126 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00074755
CAS nummer: 702-79-4
EINECS: 211-870-8
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR