1-Propanol ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

A19902.0F A19902.AP
A19902.0FEA 72.4 EUR
A19902.0F A19902.AP
1-Propanol ≥99%
1-Propanol

Purity: 99+% CAS No.: 71-23-8 Molecular Formula: CH3CH2CH2OH Formula Weight: 60.10 Flash Point: 15°(59°F) Refractive Index: 1.3850 Odour: Alcohol-like Melting Point: -127° Boiling Point: 97° Beilstein Reference: 1098242 Merck Reference: 14,7842 UN No.: 1274 MDL No.: MFCD00002941

Formule: H₃CCH₂CH₂OH
Kookpunt: 97 °C
Smeltpunt: –127 °C
Dichtheid: 0,804
Vlampunt: 15 °C (59 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002941
CAS nummer: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
Merck Index: 14,7842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR