Polyvinylpyrrolidone (K30), BioChemica®

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

A2259.1000 A2259.5000
A2259.1000EA 300 EUR
A2259.1000 A2259.5000
Polyvinylpyrrolidone (K30), BioChemica®
Polyvinylpyrrolidone
Formule: (C₆H₉NO)n
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01076626
CAS nummer: 9003-39-8

Order Now

Specification Test Results

Assay (from N) 12.0 - 12.8 %
Solubility (10 %; H2O) clear, colorless to yellowish
pH (5 %; H2O) 3.0 - 5.0
Heavy metals (as Pb) max. 0.001 %
Water (K.F.) max. 5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR