CheLuminate-HRP PicoDetect Chemiluminescent Detection Kit
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A3417.5000
A3417.5000EA 694 EUR
A3417.5000
CheLuminate-HRP PicoDetect Chemiluminescent Detection Kit
Reagentia voor elektroforese Western Blotting Reagents
The CheLuminate-HRP PicoDetect is a complete kit with ready to use reagents for chemiluminescent detection of immobilised proteins (Western blot) or immobilised nucleic acids (Southern or Northern blot), conjugated with horseradish peroxidase (HRP) directly or indirectly.
  • High sensitivity non-radioactive detection system
  • Stable hard copy results on film
  • Only small amounts of antibody required
  • Detection may be achieved in short exposure times (minutes)
  • High resolution