N-Lauroylsarcosine sodium salt 30% in aqueous solution

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate

A3457.0500
A3457.0500EA 82.5 EUR
A3457.0500
N-Lauroylsarcosine sodium salt 30% in aqueous solution
N-Lauroylsarcosine sodium salt
Formule: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042728
CAS nummer: 137-16-6
EINECS: 205-281-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=48c74917d6a93a32467317ef6c6fdbf24f3b6dc2&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR