Rubidium chloride

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Rubidiumchloride

A4240.0010 A4240.0050
A4240.0010EA 136 EUR
A4240.0010 A4240.0050
Rubidium chloride
Rubidium chloride
Formule: RbCl
Kookpunt: 1390 °C (1013 hPa)
Smeltpunt: 715 °C
Dichtheid: 2,8 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011187
CAS nummer: 7791-11-9
EINECS: 232-240-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR