PCR Mycoplasma Test Kit II
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A8994.0050 A8994.0025 A8994.0100
A8994.0050EA 880 EUR
A8994.0050 A8994.0025 A8994.0100
PCR Mycoplasma Test Kit II
Testsystemen voor microbiologie Micro-organisme testen
Lyophilised PCR mix for the detection of mycoplasma in cell culture by conventional PCR.
Meets criteria of section 2.6.7 of Ph. Eur.
Shipped at room temperature.
Leveringsinformatie: Supplied without Taq DNA polymerase. Kit contains PCR primer nucleotide mix, reaction buffer solution, water (PCR grade), positive template control, and internal control DNA.