Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 840)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

101920Q0 101920Q1
AATB101920Q0EA 371 EUR
AATB101920Q0 AATB101920Q1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 840)
Antilichamen
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: iFluor® 840
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Specificaties

antigeen symbool CD19
Environmentally Preferable
Type antilichaam Primair,CD19 antigen
Type antilichaam Primair,CD19 antigen
Clonality Monoklonaal
Conjugatie iFluor® 840
Clone HIB19
Reactivity Human
Host Mouse
Isotype
ImmunoFluorescentie Yes
Flowcytometrie Yes
Purification Affinity chromatography
Kruisadsorptie No
Opslagtemperatuur 2...8 °C with minimised light exposure. Do not freeze.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR