Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 700)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

103400J0 103400J1
AATB103400J0EA 371 EUR
AATB103400J0 AATB103400J1
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 700)
Antilichamen
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD34 antigen
Clonality: Monoclonal
Clone: 4H11
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR