Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet 510)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10340110 10340111
AATB10340110EA 371 EUR
AATB10340110 AATB10340111
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet 510)
Antilichamen
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Specificaties

antigeen symbool CD34 CD34
Environmentally Preferable
Type antilichaam CD34 antigen,Primair CD34 antigen,Primair
Type antilichaam CD34 antigen,Primair CD34 antigen,Primair
Clonality Monoklonaal Monoklonaal
Conjugatie mFluor™ Violet 510 mFluor™ Violet 510
Clone 4H11 4H11
Reactivity Human
Host Mouse Mouse
Isotype IgG1 IgG1
ImmunoFluorescentie Yes Yes
Flowcytometrie Yes Yes
Purification Affinity chromatography Affinity chromatography
Kruisadsorptie No No
Opslagtemperatuur 2...8 °C with minimised light exposure. Do not freeze. 2...8 °C with minimised light exposure. Do not freeze.
Shipping temperature 20 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR