Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/iFluor® 750)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

103801R0 103801R1 103801R2
AATB103801R0EA 319 EUR
AATB103801R0 AATB103801R1 AATB103801R2
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/iFluor® 750)
Antilichamen
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: PE/iFluor® 750
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Specificaties

antigeen symbool CD38 CD38
Environmentally Preferable
Type antilichaam Primair,CD38 antigen Primair,CD38 antigen
Type antilichaam Primair,CD38 antigen Primair,CD38 antigen
Clonality Monoklonaal Monoklonaal
Conjugatie PE (Phycoerythrin)/iFluor® 750 PE (Phycoerythrin)/iFluor® 750
Clone HIT2 HIT2
Reactivity Human
Host Mouse Mouse
Isotype IgG1 IgG1
ImmunoFluorescentie Yes Yes
Flowcytometrie Yes Yes
Purification Affinity chromatography Affinity chromatography
Kruisadsorptie No No
Opslagtemperatuur 2...8 °C with minimised light exposure. Do not freeze. 2...8 °C with minimised light exposure. Do not freeze.
Shipping temperature 20 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR