Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/Texas Red®)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

103801S0 103801S1 103801S2
AATB103801S0EA 319 EUR
AATB103801S0 AATB103801S1 AATB103801S2
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/Texas Red®)
Antilichamen
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: PE/Texas Red®
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR