Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 560)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

104410A0 104410A1
AATB104410A0EA 371 EUR
AATB104410A0 AATB104410A1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 560)
Antilichamen
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 560
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR