5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥90%

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

113 114 115
AATB113EA 152 EUR
AATB113 AATB114 AATB115
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥90%
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)

5-FAM, SE is the amine-reactive succinimidyl ester of single isomer 5-FAM acid.

Formule: C₂₅H₁₅NO₉
MDL Nummer: MFCD00077322
CAS nummer: 92557-80-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47610eeaecfe0164a88fe381d6d7a09565d50115&vid=f3c00ee395340a181740bfb53be02a7ee9e47c81&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR